Merck
  • Air-liquid interfacial self-assembly of conjugated block copolymers into ordered nanowire arrays.
Air-liquid interfacial self-assembly of conjugated block copolymers into ordered nanowire arrays.
ACS nano (2014-12-09)
Ma Helen M Cativo, David K Kim, Robert A Riggleman, Kevin G Yager, Stephen S Nonnenmann, Huikuan Chao, Dawn A Bonnell, Charles T Black, Cherie R Kagan, So-Jung Park
ZUSAMMENFASSUNG
The ability to control the molecular packing and nanoscale morphology of conjugated polymers is important for many of their applications. Here, we report the fabrication of well-ordered nanoarrays of conjugated polymers, based on the self-assembly of conjugated block copolymers at the air-liquid interface. We demonstrate that the self-assembly of poly(3-hexylthiophene)-block-poly(ethylene glycol) (P3HT-b-PEG) at the air-water interface leads to large-area free-standing films of well-aligned P3HT nanowires. Block copolymers with high P3HT contents (82-91%) formed well-ordered nanoarrays at the interface. The fluidic nature of the interface, block copolymer architecture, and rigid nature of P3HT were necessary for the formation of well-ordered nanostructures. The free-standing films formed at the interface can be readily transferred to arbitrary solid substrates. The P3HT-b-PEG films are integrated in field-effect transistors and show orders of magnitude higher charge carrier mobility than spin-cast films, demonstrating that the air-liquid interfacial self-assembly is an effective thin film fabrication tool for conjugated block copolymers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexamethyldisilazan, reagent grade, ≥99%
Sigma-Aldrich
Hexamethyldisilazan, ReagentPlus®, 99.9%
Supelco
Hexamethyldisilazan, derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Hexamethyldisilazan, Wacker Chemie AG, ≥97.0% (GC)