Merck
Alterations in the expression of p53, KLF4, and p21 in neuroendocrine lung tumors.
Archives of pathology & laboratory medicine (2014-07-01)
José Manuel Naranjo Gómez, Jose Fernando Val Bernal, Pilar García Arranz, Servando Lazuén Fernández, Jose Javier Gómez Roman
ZUSAMMENFASSUNG
Neuroendocrine lung neoplasms are a heterogeneous group of tumors with different clinical behavior and prognosis. To evaluate the expression of p53, KLF4, and p21 in neuroendocrine lung neoplasms and to analyze the influence that expression has on the prognosis of those tumors. All neuroendocrine lung neoplasms (N = 109) resected in our institution were reviewed, with the collection of histologic slides and paraffin blocks of 47 typical carcinoids (43%), 9 atypical carcinoids (8%), 35 large cell neuroendocrine carcinomas (32%), and 18 small cell lung carcinomas (17%), as well as 10 tumorlets (100%). Four tissue microarrays were performed. Follow-up was assessed in all cases (119 of 119; 100%). p53 protein immunostaining results were negative in both the tumorlets and typical carcinoids and were overexpressed in 11% (1 of 9) of the atypical carcinoids and in 68% (36 of 53) of the carcinomas. KLF4 results were positive in all tumorlets (10 of 10; 100%), 32% (15 of 47) of the typical carcinoids, 44% (4 of 9) of the atypical carcinoids, and 62% (33 of 53) of the carcinomas. p21 expression did not differ among the groups. The lack of KLF4 and p21 expression was associated with an accumulation of aggressive features in typical carcinoids (P = .04 and P = .004, respectively, Fisher exact test). p53, KLF4, and p21 showed altered expression patterns in pulmonary neuroendocrine neoplasms. Lack of KLF4 and p21 expression was associated with accumulation of aggressive features in typical carcinoids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Hämatoxylin, certified by the BSC