Merck
  • Guided bone regeneration produced by new mineralized and reticulated collagen membranes in critical-sized rat calvarial defects.
Guided bone regeneration produced by new mineralized and reticulated collagen membranes in critical-sized rat calvarial defects.
Experimental biology and medicine (Maywood, N.J.) (2014-09-24)
Denusa M Veríssimo, Renata F C Leitão, Sônia D Figueiró, Júlio C Góes, Vilma Lima, Charles O Silveira, Gerly A C Brito
ZUSAMMENFASSUNG
The aim of this study was to evaluate the bone regenerative effect of glutaraldehyde (GA) cross-linking on mineralized polyanionic collagen membranes in critical-sized defects on rat calvarias. Bone calvarial defects were induced in Wistar rats, which were then divided into five groups: a sham group; a control group, which received a commercial membrane; and GA, 25GA, and 75GA groups, which received one of three different polyanionic collagen membranes mineralized by 0, 25, or 75 hydroxyapatite cycles and then cross-linked by GA. Bone formation was evaluated based on digital radiography and computerized tomography. Histological analyses were performed 4 and 12 weeks after the surgical procedure to observe bone formation, membrane resorption, and fibrous tissue surrounding the membranes. Measurement of myeloperoxidase activity, tumor necrosis factor alpha, and interleukin 1beta production was performed 24 h after surgery. The percentage of new bone formation in the GA, 25GA, and 75GA groups was higher compared with the control and sham groups. In the GA and 25 GA groups, the membranes were still in place and were contained in a thick fibrous capsule after 12 weeks. No significant difference was found among the groups regarding myeloperoxidase activity and interleukin 1beta levels, although the GA, 25GA, and 75GA groups presented decreased levels of tumor necrosis factor alpha compared with the control group. These new GA cross-linked membranes accelerated bone healing of the calvarium defects and did not induce inflammation. In addition, unlike the control membrane, the experimental membranes were not absorbed during the analyzed period, so they may offer advantages in large bone defects where prolonged membrane barrier functions are desirable.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥96.0% (AT)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
USP
Cetrimoniumbromid, United States Pharmacopeia (USP) Reference Standard
Supelco
Hexadecyltrimethylammoniumbromid, analytical standard
Supelco
Hexadecyltrimethylammoniumbromid, suitable for ion pair chromatography, LiChropur