Merck
  • Comparative studies of N-glycans and glycosaminoglycans present in SIRC (Statens Seruminstitut rabbit cornea) cells and corneal epithelial cells from rabbit eyes.
Comparative studies of N-glycans and glycosaminoglycans present in SIRC (Statens Seruminstitut rabbit cornea) cells and corneal epithelial cells from rabbit eyes.
Current eye research (2014-01-10)
Kinya Iwatsuka, Hiroki Iwamoto, Mitsuhiro Kinoshita, Katsuhiro Inada, Shin-Ichi Yasueda, Kazuaki Kakehi
ZUSAMMENFASSUNG
We compared cultured Statens Seruminstitut rabbit cornea (SIRC) cells and corneal epithelial cells from rabbit eyes by analyzing their N-glycans and glycosaminoglycans (GAGs). This work is a fundamental study on the efficacy of using cultured cells instead of animals for drug development. N-Glycans and GAGs from SIRC cell monolayers and corneal epithelial cells of rabbit eyes were analyzed by capillary electrophoresis (CE) and a combination of high-performance liquid chromatography (HPLC) and mass spectrometry. High mannose-type glycans and a hybrid-type glycan were the common N-glycans in SIRC cells and corneal epithelial cells of rabbit eyes. Mono-fucosylated biantennary glycans with or without one N-acetylneuraminic acid residue were observed only in SIRC cells. Hyaluronic acid was the only measurable GAG in the corneal epithelial cells of rabbit eyes. In contrast, hyaluronic acid and chondroitin sulfates were abundantly present in SIRC cells. Profiles of both N-glycans and GAGs were conspicuously different between SIRC cells and corneal epithelial cells of rabbit eyes. This report will be useful for the evaluation of pharmaceutical candidates when animals or cultured cells are employed in drug development studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumcyanoborhydrid, reagent grade, 95%
Sigma-Aldrich
2,5-Dihydroxybenzoesäure, 98%
Supelco
2,5-Dihydroxybenzoesäure, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Anthranilsäure, reagent grade, ≥98%
Sigma-Aldrich
Natriumcyanoborhydrid -Lösung, 1.0 M in THF
Sigma-Aldrich
Natriumcyanoborhydrid -Lösung, 5.0 M in 1 M NaOH
Sigma-Aldrich
Anthranilsäure, puriss. p.a., ≥99.5% (T)
Supelco
2,5-Dihydroxybenzoesäure, analytical standard
Supelco
2,5-Dihydroxybenzoesäure, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure