Merck
According to the CPLL proteome sheriffs, not all aperitifs are created equal!
Biochimica et biophysica acta (2014-05-08)
María Jesús Lerma-García, Alfonsina D'Amato, Elisa Fasoli, Ernesto Francisco Simó-Alfonso, Pier Giorgio Righetti
ZUSAMMENFASSUNG
Combinatorial peptide ligand libraries (CPLLs) have been adopted for investigating the proteome of a popular aperitif in Northern Italy, called "Amaro Branzi", stated to be an infusion of a secret herbal mixture, of which some ingredients are declared on the label, namely Angelica officinalis, Gentiana lutea and orange peel, sweetened by a final addition of honey. In order to assess the genuineness of this commercial liqueur, we have prepared extracts of the three vegetable ingredients, assessed their proteomes, and compared them to the one found in the aperitif. The amaro's proteome was identified via prior capture with CPLLs at two different pH values (2.2 and 4.8). Via mass spectrometry analysis of the recovered fractions, after elution of the captured populations in 4% boiling SDS, we could confirm the presence of the following: six proteins originating from honey, 11 from orange peels, 29 from G. lutea and 46 from A. officinalis (including shared species), plus 33 species which could not be attributed to the other secret ingredients, due to paucity of genomic data on plant proteins, for a total of 93 unique gene products (merging shared proteins). This fully confirmed the genuineness of the product. Considering that most of these species could be present in trace amounts, undetectable by conventional techniques, the CPLL methodology, due to its ability to enhance the signal of trace components up to 3 to 4 orders of magnitude, could represent a powerful tool for investigating the genuineness and natural origin of commercial beverages in order to protect consumers from adulterated products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamid, analytical standard
Sigma-Aldrich
Acrylamid, purum, ≥98.0% (GC)
Supelco
Acrylamid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acrylamid -Lösung, 40% in H2O
Sigma-Aldrich
Acrylamid, for Northern and Southern blotting, powder blend