Merck
  • Impact of CYP2C9 polymorphism found in the Chinese population on the metabolism of propofol in vitro.
Impact of CYP2C9 polymorphism found in the Chinese population on the metabolism of propofol in vitro.
Biological & pharmaceutical bulletin (2015-04-04)
Qing-Quan Lian, Pei-Pei Pan, Jun-Wei Li, Han Lin, Guo-Xin Hu, Ming-Zhang Zuo, Jian-Ping Cai
ZUSAMMENFASSUNG
The microsomal CYP2C9 alleles involved in the biotransformation of propofol, a widely used anesthetic agent, were investigated in vitro. To examine the enzymatic activity of the CYP2C9 alleles, kinetic parameters for propofol 4-hydroxylation were determined in recombinant human P450s CYP2C9 microsomes from Sf21 insects cells carrying CYP2C9*1 and other variants. Some of the variants showed decreased enzyme activity compared with the wild type, as previously reported. Two variants (CYP2C9*36 and *56) were found substantially to increase intrinsic clearance relative to the wild type variant. Most variants significantly (p<0.05) decreased intrinsic clearance of propofol compared with the wild type, except *11, *47, and *54. This study is the first to report these rare alleles for propofol metabolism, providing fundamental data for further clinical studies on CYP2C9 alleles for propofol metabolism in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Propofol, European Pharmacopoeia (EP) Reference Standard
Propofol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard