Merck
  • Flow sorting and exome sequencing reveal the oncogenome of primary Hodgkin and Reed-Sternberg cells.
Flow sorting and exome sequencing reveal the oncogenome of primary Hodgkin and Reed-Sternberg cells.
Blood (2014-12-10)
Jonathan Reichel, Amy Chadburn, Paul G Rubinstein, Lisa Giulino-Roth, Wayne Tam, Yifang Liu, Rafael Gaiolla, Kenneth Eng, Joshua Brody, Giorgio Inghirami, Carmelo Carlo-Stella, Armando Santoro, Daoud Rahal, Jennifer Totonchy, Olivier Elemento, Ethel Cesarman, Mikhail Roshal
ZUSAMMENFASSUNG
Classical Hodgkin lymphoma (cHL) is characterized by sparsely distributed Hodgkin and Reed-Sternberg (HRS) cells amid reactive host background, complicating the acquisition of neoplastic DNA without extensive background contamination. We overcame this limitation by using flow-sorted HRS and intratumor T cells and optimized low-input exome sequencing of 10 patient samples to reveal alterations in genes involved in antigen presentation, chromosome integrity, transcriptional regulation, and ubiquitination. β-2-microglobulin (B2M) is the most commonly altered gene in HRS cells, with 7 of 10 cases having inactivating mutations that lead to loss of major histocompatibility complex class I (MHC-I) expression. Enforced wild-type B2M expression in a cHL cell line restored MHC-I expression. In an extended cohort of 145 patients, the absence of B2M protein in the HRS cells was associated with lower stage of disease, younger age at diagnosis, and better overall and progression-free survival. B2M-deficient cases encompassed most of the nodular sclerosis subtype cases and only a minority of mixed cellularity cases, suggesting that B2M deficiency determines the tumor microenvironment and may define a major subset of cHL that has more uniform clinical and morphologic features. In addition, we report previously unknown genetic alterations that may render selected patients sensitive to specific targeted therapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Hämatoxylin, certified by the BSC