Merck
  • Metabolism, excretion and pharmacokinetics of [14C]crizotinib following oral administration to healthy subjects.
Metabolism, excretion and pharmacokinetics of [14C]crizotinib following oral administration to healthy subjects.
Xenobiotica; the fate of foreign compounds in biological systems (2014-07-19)
Theodore R Johnson, Weiwei Tan, Lance Goulet, Evan B Smith, Shinji Yamazaki, Gregory S Walker, Melissa T O'Gorman, Gabriella Bedarida, Helen Y Zou, James G Christensen, Leslie N Nguyen, Zhongzhou Shen, Deepak Dalvie, Akintunde Bello, Bill J Smith
ZUSAMMENFASSUNG
1. Crizotinib (XALKORI®), an oral inhibitor of anaplastic lymphoma kinase (ALK) and mesenchymal-epithelial transition factor kinase (c-Met), is currently approved for the treatment of patients with non-small cell lung cancer that is ALK-positive. 2. The metabolism, excretion and pharmacokinetics of crizotinib were investigated following administration of a single oral dose of 250 mg/100 µCi [(14)C]crizotinib to six healthy male subjects. 3. Mean recovery of [(14)C]crizotinib-related radioactivity in excreta samples was 85% of the dose (63% in feces and 22% in urine). 4. Crizotinib and its metabolite, crizotinib lactam, were the major components circulating in plasma, accounting for 33% and 10%, respectively, of the 0-96 h plasma radioactivity. Unchanged crizotinib was the major excreted component in feces (∼ 53% of the dose). In urine, crizotinib and O-desalkyl crizotinib lactam accounted for ∼ 2% and 5% of the dose, respectively. Collectively, these data indicate that the primary clearance pathway for crizotinib in humans is oxidative metabolism/hepatic elimination. 5. Based on plasma exposure in healthy subjects following a single dose of crizotinib and in vitro potency against ALK and c-Met, the crizotinib lactam diastereomers are not anticipated to contribute significantly to in vivo activity; however, additional assessment in cancer patients is warranted.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Crizotinib, ≥98% (HPLC)
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.