Merck
  • CD4+ T cell-derived IL-21 and deprivation of CD40 signaling favor the in vivo development of granzyme B-expressing regulatory B cells in HIV patients.
CD4+ T cell-derived IL-21 and deprivation of CD40 signaling favor the in vivo development of granzyme B-expressing regulatory B cells in HIV patients.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-18)
Christof Kaltenmeier, Ali Gawanbacht, Thamara Beyer, Stefanie Lindner, Timo Trzaska, Johannes Andreas van der Merwe, Georg Härter, Beate Grüner, Dorit Fabricius, Ramin Lotfi, Klaus Schwarz, Catharina Schütz, Manfred Hönig, Ansgar Schulz, Peter Kern, Martin Bommer, Hubert Schrezenmeier, Frank Kirchhoff, Bernd Jahrsdörfer
ZUSAMMENFASSUNG
IL-21 can induce both plasma cells and regulatory B cells. In this article, we demonstrate that untreated HIV patients display CD4(+) T cells with enhanced IL-21 expression and high in vivo frequencies of regulatory B cells overexpressing the serine protease granzyme B. Granzyme B-expressing regulatory B cells (GraB cells) cells from HIV patients exhibit increased expression of CD5, CD43, CD86, and CD147 but do not produce IL-10. The main functional characteristic of their regulatory activity is direct granzyme B-dependent degradation of the TCR-ζ-chain, resulting in significantly decreased proliferative T cell responses. Although Th cells from HIV patients secrete IL-21 in a Nef-dependent manner, they barely express CD40L. When culturing such IL-21(+)CD40L(-) Th cells with B cells, the former directly induce B cell differentiation into GraB cells. In contrast, the addition of soluble CD40L multimers to T cell/B cell cultures redirects B cell differentiation toward plasma cells, indicating that CD40L determines the direction of IL-21-dependent B cell differentiation. As proof of principle, we confirmed this mechanism in a patient lacking intact CD40 signaling due to a NEMO mutation. The majority of peripheral B cells from this patient were GraB cells and strongly suppressed T cell proliferation. In conclusion, GraB cells represent potent regulatory B cells in humans that are phenotypically and functionally distinct from B10 cells and occur in early HIV infection. GraB cells may contribute significantly to immune dysfunction in HIV patients, and may also explain ineffective Ab responses after vaccination. The use of soluble CD40L multimers may help to improve vaccination responses in HIV patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%