Merck
Hypoxia-induced miR-497 decreases glioma cell sensitivity to TMZ by inhibiting apoptosis.
FEBS letters (2014-08-01)
Jin Lan, Yajun Xue, Huairui Chen, Sanhu Zhao, Zhijian Wu, Jun Fang, Cong Han, Meiqing Lou
ZUSAMMENFASSUNG
Understanding the resistance of glioma cells to chemotherapy has been an enormous challenge. In particular, mechanisms by which tumor cells acquire resistance to chemotherapy under hypoxic conditions are not fully understood. In this study, we have found that miR-497 is overexpressed in glioma and that hypoxia can induce the expression of miR-497 at the transcriptional level by binding with the hypoxia response element in the promoter. Ectopic overexpression of miR-497 promotes chemotherapy resistance in glioma cells by targeting PDCD4, a tumor suppressor that is involved in apoptosis. In contrast, the inhibition of miR-497 enhances apoptosis and increases the sensitivity of glioma cells to TMZ. These results suggest that miR-497 is a potential molecular target for glioma therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Temozolomid, ≥98% (HPLC)
Supelco
Temozolomid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Temozolomid, VETRANAL®, analytical standard