Merck
  • Phytochemical profiles, antioxidant activities of functional herb Abrus cantoniensis and Abrus mollis.
Phytochemical profiles, antioxidant activities of functional herb Abrus cantoniensis and Abrus mollis.
Food chemistry (2015-02-11)
Mei Yang, Qing Shen, Lin-Qiu Li, Ye-Qing Huang, Hon-Yeung Cheung
ZUSAMMENFASSUNG
It has been claimed that consumptions of Abrus cantoniensis (AC) and Abrus mollis (AM) as folk beverages and soups are good to cleanse liver toxicants and prevent liver diseases. There is scant information on the phytochemical profiles and antioxidant activities of these two varieties. Five major phytochemicals in these two cultivars were qualitatively and quantitatively compared using UPLC-PDA. A high level of total phenolic content (TPC) and total flavonoid content (TFC) was found in AC and AM. AC, in general, showed some antioxidant activities comparable to that of BHT, and stronger radical scavenging activities and higher reducing power than that of AM (p<0.05). When principal component analysis (PCA) was applied, high correlation between TPC, TFC and their antioxidant activities was found. Hence, this study proved that, both AC and AM could serve as antioxidant-rich component in foods or beverages to promote health function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Adenin, ≥99%
Supelco
Butyliertes Hydroxytoluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, ≥98%
Sigma-Aldrich
Phenazin-Methosulfat
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
Adenin, BioReagent, suitable for cell culture
Millipore
2,3,5-Triphenyl-tetrazoliumchlorid -Lösung, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
USP
Butyliertes Hydroxytoluol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Linolsäure, analytical standard
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
Phenazin, 98%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, tested according to Ph. Eur.
Supelco
Natriumnitroprussid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenin, BioReagent, suitable for plant cell culture, ≥99%
Supelco
Adenin, Pharmaceutical Secondary Standard; Certified Reference Material
Adenin, European Pharmacopoeia (EP) Reference Standard