Merck
The apolipoprotein B/apolipoprotein A-I ratio as a potential marker of plasma atherogenicity.
Disease markers (2015-04-09)
Anastasiya M Kaneva, Natalya N Potolitsyna, Evgeny R Bojko, Jon Ø Odland
ZUSAMMENFASSUNG
The apolipoprotein (apo) B/apoA-I ratio represents the balance between apoB-rich atherogenic particles and apoA-I-rich antiatherogenic particles, and this ratio is considered to be a marker of cardiovascular risk. Although many studies have demonstrated the importance of the apoB/apoA-I ratio in predicting the presence or absence of cardiovascular disease, less is known about apoB/apoA-I ratio as a marker of plasma atherogenicity. A total of 157 normolipidemic men aged 20-59 years were included in the study. The plasma levels of total cholesterol (TC), triglycerides (TG), high-density lipoprotein cholesterol (HDL-C), apoA-I, apoB, and apoE were determined after a 12 h fasting period. The median of the apoB/apoA-I ratio in the studied normolipidemic subjects was 0.52, with values ranging from 0.19 to 2.60. The percentage of subjects with the apoB/apoA-I ratio exceeding 0.9 (the accepted risk value of cardiovascular disease) was 19.1%. The subjects with apoB/apoA-I>0.9 were characterized by higher TG levels and atherogenic index of plasma (AIP) and lower values of ratio of low-density lipoprotein cholesterol (LDL-C) to apoB (LDL-C/apoB) and apoE levels compared with men with apoB/apoA-I<0.9. Despite normolipidemia, the subjects with the unfavorable apoB/apoA-I ratio had more atherogenic lipid profile.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumisopropoxid, ≥98%
Sigma-Aldrich
Aluminiumisopropoxid, ≥99.99% trace metals basis
Sigma-Aldrich
Autocamtide 2-related inhibitory peptide, ≥97% (HPLC), lyophilized powder