Merck
  • Anti-HBV activity of the different extracts from Phyllanthus rheedei Wight in cell culture based assay systems.
Anti-HBV activity of the different extracts from Phyllanthus rheedei Wight in cell culture based assay systems.
Journal of ethnopharmacology (2014-09-16)
V Suresh, Jose Sojan, N Krishna Radhika, V V Asha
ZUSAMMENFASSUNG
Phyllanthusrheedei Wight is a plant used by Muthuvan tribes of Kerala for treating liver related diseases. The different extracts of Phyllanthus rheedei were analysed on cell lines were viz, PLC/PRF, Hep3B, FLCII10 and HepG2215 for its anti-HBV property. The analysis was done through ELISA, SQRT-PCR and immuno blotting. The most active extract was then divided in to fractions using HPTLC and the most active fraction was further identified. From the screening experiments it was shown that the ethanol extract of this plant has the maximum activity in lowering the viral markers like HBsAg, HBV Core and HBV X protein and whole virions with comparatively lesser cytotoxicity. The dose responses of this particular extract were further established. This study concluded that the ethanol extract of Phyllanthusrheedei is very much effective in preventing the multiplication of HBV at the cellular level. This study scientifically validated the tribal claim of the use of this plant for severe liver disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Ethylacetat, analytical standard
Sigma-Aldrich
Ellagsäure, ≥95% (HPLC), powder, from tree bark
Supelco
Toluol, analytical standard
Supelco
Ethylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Supelco
Ellagsäure, analytical standard
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%