Merck
  • An orally administrated nucleotide-delivery vehicle targeting colonic macrophages for the treatment of inflammatory bowel disease.
An orally administrated nucleotide-delivery vehicle targeting colonic macrophages for the treatment of inflammatory bowel disease.
Biomaterials (2015-02-24)
Zhen Huang, Jingjing Gan, Lixin Jia, Guangxing Guo, Chunming Wang, Yuhui Zang, Zhi Ding, Jiangning Chen, Junfeng Zhang, Lei Dong
ZUSAMMENFASSUNG
Tumor necrosis factor-alpha (TNF-α) plays a central role in the pathogenesis of inflammatory bowel disease (IBD). Anti-TNF-α therapies have shown protective effects against colitis, but an efficient tool for target suppression of its secretion - ideally via oral administration - remains in urgent demand. In the colon tissue, TNF-α is mainly secreted by the colonic macrophages. Here, we report an orally-administrated microspheric vehicle that can target the colonic macrophages and suppress the local expression of TNF-α for IBD treatment. This vehicle is formed by cationic konjac glucomannan (cKGM), phytagel and an antisense oligonucleotide against TNF-α. It was given to dextran sodium sulfate (DSS) colitic mice via gastric perfusion. The unique swelling properties of cKGM enabled the spontaneous release of cKGM& antisense nucleotide (ASO) nano-complex from the phytagel scaffold into the colon lumen, where the ASO was transferred into colonic macrophages via receptor-mediated phagocytosis. The treatment significantly decreased the local level of TNF-α and alleviated the symptoms of colitis in the mice. In summary, our study demonstrates a convenient, orally-administrated drug delivery system that effectively targets colonic macrophages for suppression of TNF-α expression. It may represent a promising therapeutic approach in the treatment of IBD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumsulfat, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder
Millipore
D-(+)-Mannose, ≥99%, suitable for microbiology
Sigma-Aldrich
Natriumsulfat, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
2-Phenylindol, technical grade, 95%
Sigma-Aldrich
Natriumsulfat, ≥99.0%, suitable for plant cell culture
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology