Merck
  • Comparative analysis of the treatment of chronic antipsychotic drugs on epileptic susceptibility in genetically epilepsy-prone rats.
Comparative analysis of the treatment of chronic antipsychotic drugs on epileptic susceptibility in genetically epilepsy-prone rats.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics (2014-11-19)
Rita Citraro, Antonio Leo, Rossana Aiello, Michela Pugliese, Emilio Russo, Giovambattista De Sarro
ZUSAMMENFASSUNG
Antipsychotic drugs (APs) are of great benefit in several psychiatric disorders, but they can be associated with various adverse effects, including seizures. To investigate the effects of chronic antipsychotic treatment on seizure susceptibility in genetically epilepsy-prone rats, some APs were administered for 7 weeks, and seizure susceptibility (audiogenic seizures) was evaluated once a week during treatment and for 5 weeks after drug withdrawal. Furthermore, acute and subchronic (5-day treatment) effects were also measured. Rats received haloperidol (0.2-1.0 mg/kg), clozapine (1-5 mg/kg), risperidone (0.03-0.50 mg/kg), quetiapine (2-10 mg/kg), aripriprazole (0.2-1.0 mg/kg), and olanzapine (0.13-0.66 mg/kg), and tested according to treatment duration. Acute administration of APs had no effect on seizures, whereas, after regular treatment, aripiprazole reduced seizure severity; haloperidol had no effects and all other APs increased seizure severity. In chronically treated rats, clozapine showed the most marked proconvulsant effects, followed by risperidone and olanzapine. Quetiapine and haloperidol had only modest effects, and aripiprazole was anticonvulsant. Finally, the proconvulsant effects lasted at least 2-3 weeks after treatment suspension; for aripiprazole, a proconvulsant rebound effect was observed. Taken together, these results indicate and confirm that APs might have the potential to increase the severity of audiogenic seizures but that aripiprazole may exert anticonvulsant effects. The use of APs in patients, particularly in patients with epilepsy, should be monitored for seizure occurrence, including during the time after cessation of therapy. Further studies will determine whether aripiprazole really has a potential as an anticonvulsant drug and might also be clinically relevant for epileptic patients with psychiatric comorbidities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Clozapin
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Risperidon, ≥98% (HPLC), powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Olanzapin, ≥98% (HPLC)
USP
Olanzapin, United States Pharmacopeia (USP) Reference Standard
USP
Risperidon, United States Pharmacopeia (USP) Reference Standard
USP
Clozapin-Auflösungsmischung, United States Pharmacopeia (USP) Reference Standard
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Clozapin, European Pharmacopoeia (EP) Reference Standard
USP
Clozapin, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-on, VETRANAL®, analytical standard
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Olanzapin, European Pharmacopoeia (EP) Reference Standard
Risperidon für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Risperidon, European Pharmacopoeia (EP) Reference Standard
Millipore
Bifido Selektives Supplement B, suitable for microbiology
Olanzapin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Clozapin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard