Merck
  • The palladacycle, AJ-5, exhibits anti-tumour and anti-cancer stem cell activity in breast cancer cells.
The palladacycle, AJ-5, exhibits anti-tumour and anti-cancer stem cell activity in breast cancer cells.
Cancer letters (2014-12-03)
Saeb Aliwaini, Jade Peres, Wendy L Kröger, Angelique Blanckenberg, Jo de la Mare, Adrienne L Edkins, Selwyn Mapolie, Sharon Prince
ZUSAMMENFASSUNG
Breast cancer is the most common malignancy amongst women worldwide but despite enormous efforts to address this problem, there is still limited success with most of the current therapeutic strategies. The current study describes the anti-cancer activity of a binuclear palladacycle complex (AJ-5) in oestrogen receptor positive (MCF7) and oestrogen receptor negative (MDA-MB-231) breast cancer cells as well as human breast cancer stem cells. AJ-5 is shown to induce DNA double strand breaks leading to intrinsic and extrinsic apoptosis and autophagy cell death pathways which are mediated by the p38 MAP kinase. This study provides evidence that AJ-5 is potentially an effective compound in the treatment of breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Wortmannin aus Penicillium funiculosum, from Penicillium funiculosum, ≥98% (HPLC and TLC)