Merck
  • Knockdown of T-bet expression in Mart-127-35 -specific T-cell-receptor-engineered human CD4(+)  CD25(-) and CD8(+) T cells attenuates effector function.
Knockdown of T-bet expression in Mart-127-35 -specific T-cell-receptor-engineered human CD4(+)  CD25(-) and CD8(+) T cells attenuates effector function.
Immunology (2014-12-17)
Sidharth S Jha, Nitya G Chakraborty, Prashant Singh, Bijay Mukherji, David I Dorsky
ZUSAMMENFASSUNG
Gene transfer to create tumour epitope-specific cytolytic T cells for adoptive immunotherapy of cancer remains an area of active inquiry. When the Mart-127-35 -specific DMF5 T-cell receptor (TCR) is transferred into peripheral human CD4(+) T cells, the reprogrammed cells exhibit a T helper type 1 (Th1) phenotype with significant multifactorial effector capabilities. The T-bet transcription factor plays an important role in determination of the Th1 differentiation pathway. To gain a deeper understanding of how T-bet controls the outcome of human T-cell reprogramming by gene transfer, we developed a system for examining the effects of short hairpin RNA-mediated T-bet gene knockdown in sorted cell populations uniformly expressing the knockdown construct. In this system, using activated peripheral human CD4(+)  CD25(-) and CD8(+) T cells, T-bet knockdown led to attenuation of the interferon-γ response to both antigen-specific and non-specific TCR stimulation. The interleukin-2 (IL-2) antigen-specific response was not attenuated by T-bet knockdown. Also, in TCR-reprogrammed CD8(+) cells, the cytolytic effector response was attenuated by T-bet knockdown. T-bet knockdown did not cause redirection into a Th2 differentiation pathway, and no increased IL-4, IL-10, or IL-17 response was detected in this system. These results indicate that T-bet expression is required for maintenance of the CD4(+)  CD25(-) and CD8(+) effector phenotypes in TCR-reprogrammed human T cells. They also suggest that the activation protocol necessary for transduction with retrovectors and lentivectors may commit the reprogrammed cells to the Th1 phenotype, which cannot be altered by T-bet knockdown but that there is, nevertheless, a continuous requirement of T-bet expression for interferon-γ gene activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Humanes Interleukin-2 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2, recombinant, expressed in E. coli, ~10000 U/mL