Merck
  • Improvement in spatial memory dysfunction by telmisartan through reduction of brain angiotensin II and oxidative stress in experimental uremic mice.
Improvement in spatial memory dysfunction by telmisartan through reduction of brain angiotensin II and oxidative stress in experimental uremic mice.
Life sciences (2014-08-12)
Naoki Haruyama, Kiichiro Fujisaki, Mayumi Yamato, Masahiro Eriguchi, Hideko Noguchi, Kumiko Torisu, Kazuhiko Tsuruya, Takanari Kitazono
ZUSAMMENFASSUNG
We previously reported that chronic uremia induces spatial working memory dysfunction in mice, and that it is attributed to cerebral oxidative stress. The source of oxidative stress was considered to be uremic toxins, but this remains unclear. In the present study, we examined whether the brain renin-angiotensin system was activated in the CKD mouse model, and whether it contributed to cognitive impairment. CKD was induced in 8-week-old male mice by 5/6 nephrectomy. Mice were divided into four groups: control mice administered tap water (Cont-V), control mice treated with 0.5mg/kg/day telmisartan, an angiotensin II (AII) receptor blocker, for 8 weeks (Cont-T), CKD mice administered tap water (CKD-V), and CKD mice treated with 0.5 mg/kg/day telmisartan for 8 weeks (CKD-T). After the treatment period, a radial arm water maze (RAWM) test was performed, and angiotensin II (AII) concentrations and markers of oxidative stress were measured in the brains of mice. Errors in the RAWM test were more frequent in the CKD-V group than in the Cont-V group. In addition, errors in the CKD-T group were comparable to control mice. Tissue brain AII concentrations were greater in the CKD-V group compared with the other groups. Oxidative DNA damage and lipid peroxidation in the brain were also greater in the CKD-V group compared with the other groups. Our results suggest that brain AII levels were exaggerated in CKD mice, and that this contributes to cognitive impairment through oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%