Merck
  • Electrospun chitosan-based nanocomposite mats reinforced with chitin nanocrystals for wound dressing.
Electrospun chitosan-based nanocomposite mats reinforced with chitin nanocrystals for wound dressing.
Carbohydrate polymers (2014-05-13)
Narges Naseri, Constance Algan, Valencia Jacobs, Maya John, Kristiina Oksman, Aji P Mathew
ZUSAMMENFASSUNG
The aim of this study was to develop electrospun chitosan/polyethylene oxide-based randomly oriented fiber mats reinforced with chitin nanocrystals (ChNC) for wound dressing. Microscopy studies showed porous mats of smooth and beadless fibers with diameters between 223 and 966 nm. The addition of chitin nanocrystals as well as crosslinking had a positive impact on the mechanical properties of the mats, and the crosslinked nanocomposite mats with a tensile strength of 64.9 MPa and modulus of 10.2 GPa were considered the best candidate for wound dressing application. The high surface area of the mats (35 m(2)g(-1)) was also considered beneficial for wound healing. The water vapor transmission rate of the prepared mats was between 1290 and 1,548 gm(-2)day(-1), and was in the range for injured skin or wounds. The electrospun fiber mats showed compatibility toward adipose derived stem cells, further confirming their potential use as wound dressing materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysin-hydrochlorid -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
5α-Androstan-17β-ol-3-on, VETRANAL®, analytical standard
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Millipore
Bifido Selektives Supplement B, suitable for microbiology