Merck
Regulation of neutrophil phagocytosis of Escherichia coli by antipsychotic drugs.
International immunopharmacology (2014-12-03)
Mao-Liang Chen, Semon Wu, Tzung-Chieh Tsai, Lu-Kai Wang, Fu-Ming Tsai
ZUSAMMENFASSUNG
Antipsychotic drugs (APDs) have been used to ease the symptoms of schizophrenia. APDs have recently been reported to regulate the immune response. Our previous studies revealed that the atypical APDs risperidone and clozapine and the typical APD haloperidol can inhibit the phagocytic ability of macrophages. Our research next determined the effects of APDs on the phagocytic ability of neutrophils, which are the most abundant type of white blood cells in mammals. Here we provide evidence that clozapine and haloperidol can induce increased phagocytic uptake of Escherichia coli by differentiated HL-60 cells and by purified human neutrophils. Furthermore, clozapine and haloperidol can increase the myeloperoxidase activity and IL-8 production in neutrophils. Our results also show that clozapine can inhibit E. coli survival within differentiated HL-60 cells. Furthermore, clozapine and haloperidol are shown to enhance cell surface Mac-1 expression and the activated AKT signaling pathway in purified neutrophils exposed to E. coli. These results indicate that clozapine and haloperidol can increase the phagocytic ability of neutrophils by increasing AKT activation when cells are exposed to bacteria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Clozapin
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
USP
Clozapin-Auflösungsmischung, United States Pharmacopeia (USP) Reference Standard
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Clozapin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
USP
Clozapin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Clozapin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Haloperidol für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Haloperidol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard