Merck
Optimization of pilocarpine-mediated seizure induction in immunodeficient NodScid mice.
Epilepsy research (2014-12-20)
Amanda Leung, Sandra Ahn, George Savvidis, Yeachan Kim, Danielle Iskandar, Maria Jose Luna, Kwang-Soo Kim, Miles Cunningham, Sangmi Chung
ZUSAMMENFASSUNG
Temporal lobe epilepsy (TLE) has been modeled in mice using pilocarpine induction, with variable results depending on specific strains. To allow efficient xenotransplantation for the purpose of optimizing potential cell-based therapy of human TLE, we have determined the optimal dosing strategy to produce spontaneous recurring seizures in immunodeficient NodScid mice. Multiple 100mg/kg injections of pilocarpine have been shown to be more effective than single 300-400mg/kg injections for inducing spontaneous seizures in NodScid mice. Under our optimal conditions, 88.1 ± 2.9% of the mice experienced status epilepticus (SE) with a survival rate of 61.8 ± 5.9%. Surviving SE mice displayed spontaneous recurrent seizures at a frequency of 2.8 ± 0.9 seizures/day for a duration of 41.1 ± 3.5s. The widely used method of a single injection of pilocarpine was significantly less efficient in inducing seizures in NodScid mice. Therefore, we have determined that a multiple injection "ramping up" of 100mg/kg of pilocarpine is optimal for inducing TLE-like spontaneous seizures in NodScid mice. Using this method, mice with SE efficiently developed SRS and expressed mossy fiber sprouting, a signature histopathological feature of TLE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Isofluran, European Pharmacopoeia (EP) Reference Standard