Merck
  • Coronary artery bypass grafting-related bleeding complications in real-life acute coronary syndrome patients treated with clopidogrel or ticagrelor.
Coronary artery bypass grafting-related bleeding complications in real-life acute coronary syndrome patients treated with clopidogrel or ticagrelor.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery (2014-02-01)
Emma C Hansson, Helena Rexius, Mikael Dellborg, Per Albertsson, Anders Jeppsson
ZUSAMMENFASSUNG
Ticagrelor reduces thrombotic events compared with clopidogrel in patients with acute coronary syndrome, but may also increase bleeding complications. Coronary artery bypass grafting (CABG)-related bleeding complications have not previously been compared in clopidogrel and ticagrelor-treated patients outside the controlled environment of clinical trials. Four hundred and five consecutive CABG patients with acute coronary syndrome were included in a prospective observational study. The patients were treated with aspirin and ticagrelor (n = 173) or aspirin and clopidogrel (n = 232). Ticagrelor/clopidogrel was discontinued 5 days before surgery whenever deemed possible. Major bleeding complications according to modified blood conservation using antifibrinolytics in a randomized trial criteria (postoperative blood loss >1500 ml/12 h, re-exploration, red blood cell transfusion >10 units or death because of bleeding) were compared in all patients and when ticagrelor/clopidogrel was discontinued ≥5 days (n = 280), 2-4 days (n = 40) or 0-1 day before surgery (n = 85). Major bleeding complications did not differ significantly between ticagrelor- and clopidogrel-treated patients when all patients were compared (14.5 vs 13.8%, P = 0.89). Likewise, there were no significant differences between ticagrelor and clopidogrel when either drug was discontinued ≥5 days before surgery (6.8 vs 9.9%, P = 0.40) or 2-4 days before surgery (6.3 vs 25.0%, P = 0.21). When ticagrelor/clopidogrel was discontinued 0-1 day before surgery, there was a strong trend towards higher incidence of major bleeding in ticagrelor-treated patients (41.0 vs 21.7%, P = 0.063). There was no difference in major bleeding complications overall or when ticagrelor or clopidogrel was used in accordance with guidelines. In patients on dual antiplatelet medication up to 1 day before surgery, there tended to be more bleeding complications in ticagrelor-treated patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination