Merck
  • New approach to evaluate the lubrication process in various granule filling levels and rotating mixer sizes using a thermal effusivity sensor.
New approach to evaluate the lubrication process in various granule filling levels and rotating mixer sizes using a thermal effusivity sensor.
Chemical & pharmaceutical bulletin (2015-03-12)
Jumpei Uchiyama, Shigeru Aoki, Yoshifumi Uemoto
ZUSAMMENFASSUNG
The principles of thermal effusivity are applied to an understanding of the detailed mechanisms of the lubrication process in a rotating mixer. The relationships and impact of the lubrication process by the pattern of powder flow, the filling level, and the rotating mixer size were investigated. Thermal effusivity profiles of the lubrication process, as obtained, indicate that lubrication is a two-phase process. The intersection point of the first and second phases (IPFS) is influenced by changing the filling level, thus changing the resulting number of avalanche flows created. The slope of the second phase (SSP) is influenced by the relationship between the number and the length of avalanche flows. Understanding this difference between the first and second phases is important to successfully evaluate the impact of proposed changes in the lubrication process. From this knowledge, a predictive model of the lubrication profile can be generated to allow an evaluation of proposed changes to the lubrication process. This model allows estimation of the lubrication profile at different filling levels and in different rotating mixer sizes. In this study, the actual lubrication profile almost coincides with the model predicted lubrication profile. Based on these findings, it is assumed that lubrication profiles at a commercial scale can be predicted from data generated at the laboratory scale. Further, it is assumed that changes in the filling level can also be estimated from the laboratory or current data.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Magnesiumstearat, technical grade
Sigma-Aldrich
Magnesiumstearat, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)