Merck
  • Synbiotic Lactobacillus acidophilus NCFM and cellobiose does not affect human gut bacterial diversity but increases abundance of lactobacilli, bifidobacteria and branched-chain fatty acids: a randomized, double-blinded cross-over trial.
Synbiotic Lactobacillus acidophilus NCFM and cellobiose does not affect human gut bacterial diversity but increases abundance of lactobacilli, bifidobacteria and branched-chain fatty acids: a randomized, double-blinded cross-over trial.
FEMS microbiology ecology (2014-08-08)
Gabriella C van Zanten, Lukasz Krych, Henna Röytiö, Sofia Forssten, Sampo J Lahtinen, Waleed Abu Al-Soud, Søren Sørensen, Birte Svensson, Lene Jespersen, Mogens Jakobsen
ZUSAMMENFASSUNG
Probiotics, prebiotics, and combinations thereof, that is synbiotics, have been reported to modulate gut microbiota of humans. In this study, effects of a novel synbiotic on the composition and metabolic activity of human gut microbiota were investigated. Healthy volunteers (n = 18) were enrolled in a double-blinded, randomized, and placebo-controlled cross-over study and received synbiotic [Lactobacillus acidophilus NCFM (10(9) CFU) and cellobiose (5 g)] or placebo daily for 3 weeks. Fecal samples were collected and lactobacilli numbers were quantified by qPCR. Furthermore, 454 tag-encoded amplicon pyrosequencing was used to monitor the effect of synbiotic on the composition of the microbiota. The synbiotic increased levels of Lactobacillus spp. and relative abundances of the genera Bifidobacterium, Collinsella, and Eubacterium while the genus Dialister was decreased (P < 0.05). No other effects were found on microbiota composition. Remarkably, however, the synbiotic increased concentrations of branched-chain fatty acids, measured by gas chromatography, while short-chain fatty acids were not affected.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Pivalinsäure, 99%
Sigma-Aldrich
Oxalsäure, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalsäure, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
Oxalsäure-Konzentrat, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC