Merck
MicroRNA-378 regulates adiponectin expression in adipose tissue: a new plausible mechanism.
PloS one (2014-11-08)
Masayoshi Ishida, Michio Shimabukuro, Shusuke Yagi, Sachiko Nishimoto, Chisayo Kozuka, Daiju Fukuda, Takeshi Soeki, Hiroaki Masuzaki, Masato Tsutsui, Masataka Sata
ZUSAMMENFASSUNG
Mechanisms regulating adiponectin expression have not been fully clarified. MicroRNAs (miRNAs), small non-coding RNAs that regulate gene expression, are involved in biological processes, including obesity and insulin resistance. We evaluated whether the miRNA-378 pathway is involved in regulating adiponectin expression. First, we determined a putative target site for miRNA-378 in the 3 prime untranslated region (3'UTR) of the adiponectin gene by in silico analysis. The levels of adiponectin mRNA and protein were decreased in 3T3-L1 cells overexpressing the mimic of miRNA-378. Luminescence activity in HEK293T cells expressing a renilla-luciferase-adiponectin-3'UTR sequence was inhibited by overexpressing the mimic of miRNA-378, and the decrease was reversed by adding the inhibitor of miRNA-378. Moreover, we confirmed the inhibitory effects of the mimic were cancelled in a deleted mutant of the miR-378 3'-UTR binding site. Addition of tumor necrosis factor-α (TNFα) led a upregulation of miR-378 and downregulation of adiponectin at mRNA and protein levels in 3T3-L1 cells. Level of miR-378 was higher and mRNA level of adiponectin was lower in diabetic ob/ob mice than those of normal C57BL/6 mice and levels of miR378 and adiponectin were negatively well correlated (r = -0.624, p = 0.004). We found that levels of miRNA-378 could modulate adiponectin expression via the 3'UTR sequence-binding site. Our findings warrant further investigations into the role of miRNAs in regulating the adiponectin expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tumornekrosefaktor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Tumornekrosefaktor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture