Merck
  • Neonatal uterine and vaginal cell proliferation and adenogenesis are independent of estrogen receptor 1 (ESR1) in the mouse.
Neonatal uterine and vaginal cell proliferation and adenogenesis are independent of estrogen receptor 1 (ESR1) in the mouse.
Biology of reproduction (2015-02-06)
Manjunatha K Nanjappa, Theresa I Medrano, Amelia G March, Paul S Cooke
ZUSAMMENFASSUNG
Neonatal uterus and vagina express estrogen receptor 1 (ESR1) and respond mitogenically to exogenous estrogens. However, neonatal ovariectomy does not inhibit preweaning uterine cell proliferation, indicating that this process is estrogen independent. Extensive literature suggests that ESR1 can be activated by growth factors in a ligand-independent manner and drive uterine cell proliferation. Alternatively, neonatal uterine cell proliferation could be ESR1 independent despite its obligatory role in adult luminal epithelial proliferation. To determine ESR1's role in uterine and vaginal development, we analyzed cell proliferation, apoptosis, and uterine gland development (adenogenesis) in wild-type (WT) and Esr1 knockout (Esr1KO) mice from Postnatal Day 2 to Postnatal Day 60. Uterine and vaginal cell proliferation, apoptosis, and uterine adenogenesis were comparable in WT and Esr1KO mice before weaning. By Days 29-60, glands had regressed, and uterine cell proliferation was reduced in Esr1KO mice in contrast to continued adenogenesis and proliferation in WT. Apoptosis in Esr1KO uterine epithelium was not increased compared to WT at any age, indicating that differences in cell proliferation, rather than apoptosis, cause divergence of uterine size in these two groups at puberty. Similarly, vaginal epithelial proliferation was reduced, and the epithelium became atrophic in Esr1KO mice by 29 days of age and later in Esr1KO mice. These results indicate that preweaning uterine and vaginal development is ESR1 independent but becomes dependent on ESR1 by Day 29 on. It is not yet clear what mechanisms drive preweaning vaginal and uterine development, but ligand-independent activation of ESR1 is not involved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Hämatoxylin, certified by the BSC