Merck
  • Low frequency magnetic force augments hepatic differentiation of mesenchymal stem cells on a biomagnetic nanofibrous scaffold.
Low frequency magnetic force augments hepatic differentiation of mesenchymal stem cells on a biomagnetic nanofibrous scaffold.
Journal of materials science. Materials in medicine (2014-07-12)
Dillip Kumar Bishi, Soma Guhathakurta, Jayarama Reddy Venugopal, Kotturathu Mammen Cherian, Seeram Ramakrishna
ZUSAMMENFASSUNG
Liver tissue engineering using polymeric nanofibrous scaffold and stem cells holds great promises for treating end-stage liver failures. The aim of this study was to evaluate hepatic trans-differentiation potential of human mesenchymal stem cells (hMSCs) on a biomagnetic electrospun nanofibrous scaffold fabricated from a blend of poly-L-lactide (PLLA), collagen and fibrin-rich blood clot, under the influence of a low frequency magnetic field. The scaffold was characterized for surface properties, biochemical and biomechanical parameters and bio-magnetic behaviour. Cell proliferation assay revealed that the scaffold was suitable for hMSCs adhesion and proliferation. Hepatic trans-differentiation potential of hMSCs was augmented on nanofibrous scaffold in magnetic field exposure group compared to control groups, as evident by strong expression of hepatocyte specific markers, albumin release, urea synthesis and presence of an inducible cytochrome P450 system. Our results conclude that biomagnetic scaffold of PLLA/collagen/blood clot augments hepatic trans-differentiation of hMSCs under magnetic field influence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Supelco
1,1,1,3,3,3-Hexafluor-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Harnstoff, analytical standard
Sigma-Aldrich
Harnstoff, meets USP testing specifications
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
USP
Harnstoff, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.