Merck
2, 3-dimercaptosuccinic acid-modified iron oxide clusters for magnetic resonance imaging.
Journal of pharmaceutical sciences (2014-10-23)
Fei Xiong, Caiyun Yan, Jilai Tian, Kunkun Geng, Ziyi Zhu, Lina Song, Yu Zhang, Matthew Mulvale, Ning Gu
ZUSAMMENFASSUNG
Over the last decade, various magnetic nanomaterials have been developed as magnetic resonance imaging (MRI) contrast agents; the greatest challenges encountered for clinical application have been insufficient stability. In this paper, a lyophilization method for 2, 3-dimercaptosuccinic acid-modified iron oxide (γ-Fe2 O3 @DMSA) nanoparticles was developed to simultaneously overcome two disadvantages; these include insufficient stability and low-magnetic response. After lyophilization, the clusters of γ-Fe2 O3 @DMSA with the size of 156.7 ± 15.3 nm were formed, and the stability of the lyophilized powder (γ-Fe2 O3 @DMSA-LP) increased up to over 3 years. It was also found that rehydrated γ-Fe2 O3 @DMSA-LP could be ingested by RAW264.7 cells in very large quantities. Results of pharmacokinetics and biodistribution studies in vivo indicated that γ-Fe2 O3 @DMSA-LP is a promising liver-targeted material. Furthermore, it also exhibited higher MRI efficiency and longer imaging time in the liver than the well-known product Feridex(®) . Moreover, results of vascular irritation and long-term toxicity experiments demonstrated γ-Fe2 O3 @DMSA-LP could be a nontoxic, biocompatible contrast agent in vivo. Therefore, the proposed γ-Fe2 O3 @DMSA-LP can be used as a potential MRI contrast agent in clinic for hepatic diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
1-Vinyl-2-pyrrolidon, contains sodium hydroxide as inhibitor, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
meso-2,3-Dimercaptobernsteinsäure, ~98%
Sigma-Aldrich
Glycin, 99%, FCC
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Maltose -Lösung, BioReagent, ~20% in H2O, Molecular Biology
Leucin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Leucin, ≥99% (HPLC)
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Glycin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysin-hydrochlorid -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.