Merck
  • CdSe-graphene oxide light-harvesting assembly: size-dependent electron transfer and light energy conversion aspects.
CdSe-graphene oxide light-harvesting assembly: size-dependent electron transfer and light energy conversion aspects.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-03-20)
Sachidananda Krishnamurthy, Prashant V Kamat
ZUSAMMENFASSUNG
Excited-state interaction between CdSe quantum dots (QDs) of different sizes (2.3, 3.2, and 4.2 nm diameter) and graphene oxide (GO) was probed by depositing them as films on conducting glass electrodes. The emission of smaller CdSe QDs (2.3 nm) was quenched by GO three times faster than that of larger QDs (4.2 nm). Electrophoretic deposition allowed us to sequentially deposit single or multiple layers of different sized QDs and GO assemblies on conducting glass electrodes and to modulate the photoresponse in photoelectrochemical solar cells. Superior photoconversion efficiency through the incorporation of GO was attributed to improved charge separation in the composite assembly.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Trioctylphosphin, 97%
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Trioctylphosphin, technical grade, 90%
Supelco
Schwefelsäurekonzentrat, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Kaliumpermanganat, 97%
Supelco
Schwefelsäure, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Tetradecylphosphonsäure, 98%
Sigma-Aldrich
Schwefelsäure, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Kaliumpermanganat, meets USP testing specifications