Merck
  • Investigation of the stabilizing effects of hydroxyethyl cellulose on LDH during freeze drying and freeze thawing cycles.
Investigation of the stabilizing effects of hydroxyethyl cellulose on LDH during freeze drying and freeze thawing cycles.
Pharmaceutical development and technology (2013-11-30)
Anas Al-Hussein, Henning Gieseler
ZUSAMMENFASSUNG
The objective of this study was to investigate both the cryoprotective and lyoprotective effects of the polymer hydroxyethyl cellulose (HEC) on the model protein lactate dehydrogenase (LDH) during freeze thawing and freeze drying cycles. The effect of annealing on both protein stability and the physical state of HEC was evaluated. HEC was used as a sole excipient in the protein formulations, and its stabilizing was compared to that of other excipients which are commonly used in freeze dried protein formulations. Furthermore, other quality aspects of the freeze dried samples containing solely HEC were investigated, such as, reconstitution time and product elegance. Protein stability was evaluated functionally by measuring the activity recovery of the model protein LDH. The physical state of HEC after freeze drying was investigated and compared to this of other studied solutes using differential scanning calorimetry and X-ray powder diffractometry. HEC showed superior cryoprotective effects on LDH during freeze thawing, and considerable lyoprotective effects during the freeze drying process. Annealing had limited influence on the stabilizing effect of HEC. The extensive reconstitution times of the HEC lyophilisates could be greatly improved by incorporation of the surfactant Tween 80 into the formulations prior to freeze drying.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Ammoniumsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Ammoniumsulfat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
L-Histidin, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Ammoniumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Histidin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidin, ReagentPlus®, ≥99% (TLC)
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent