Merck
  • An activated unfolded protein response promotes retinal degeneration and triggers an inflammatory response in the mouse retina.
An activated unfolded protein response promotes retinal degeneration and triggers an inflammatory response in the mouse retina.
Cell death & disease (2014-12-19)
T Rana, V M Shinde, C R Starr, A A Kruglov, E R Boitet, P Kotla, S Zolotukhin, A K Gross, M S Gorbatyuk
ZUSAMMENFASSUNG
Recent studies on the endoplasmic reticulum stress have shown that the unfolded protein response (UPR) is involved in the pathogenesis of inherited retinal degeneration caused by mutant rhodopsin. However, the main question of whether UPR activation actually triggers retinal degeneration remains to be addressed. Thus, in this study, we created a mouse model for retinal degeneration caused by a persistently activated UPR to assess the physiological and morphological parameters associated with this disease state and to highlight a potential mechanism by which the UPR can promote retinal degeneration. We performed an intraocular injection in C57BL6 mice with a known unfolded protein response (UPR) inducer, tunicamycin (Tn) and examined animals by electroretinography (ERG), spectral domain optical coherence tomography (SD-OCT) and histological analyses. We detected a significant loss of photoreceptor function (over 60%) and retinal structure (35%) 30 days post treatment. Analysis of retinal protein extracts demonstrated a significant upregulation of inflammatory markers including interleukin-1β (IL-1β), IL-6, tumor necrosis factor-α (TNF-α), monocyte chemoattractant protein-1 (MCP-1) and IBA1. Similarly, we detected a strong inflammatory response in mice expressing either Ter349Glu or T17M rhodopsin (RHO). These mutant rhodopsin species induce severe retinal degeneration and T17M rhodopsin elicits UPR activation when expressed in mice. RNA and protein analysis revealed a significant upregulation of pro- and anti-inflammatory markers such as IL-1β, IL-6, p65 nuclear factor kappa B (NF-kB) and MCP-1, as well as activation of F4/80 and IBA1 microglial markers in both the retinas expressing mutant rhodopsins. We then assessed if the Tn-induced inflammatory marker IL-1β was capable of inducing retinal degeneration by injecting C57BL6 mice with a recombinant IL-1β. We observed ~19% reduction in ERG a-wave amplitudes and a 29% loss of photoreceptor cells compared with control retinas, suggesting a potential link between pro-inflammatory cytokines and retinal pathophysiological effects. Our work demonstrates that in the context of an established animal model for ocular disease, the persistent activation of the UPR could be responsible for promoting retinal degeneration via the UPR-induced pro-inflammatory cytokine IL-1β.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tunicamycin aus Streptomyces sp.
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
L-Glutaminsäure Mononatriumsalz Hydrat, ≥99% (HPLC), powder
Supelco
Phenylephrin-Hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutaminsäure Mononatriumsalz Hydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
(R)-(−)-Phenylephrin -hydrochlorid, powder
USP
Phenylephrin -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-Kaninchen-IgG(H+L)-Antikörper der Ziege, FITC-Konjugat, Upstate®, from goat
Phenylephrin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutaminsäure Mononatriumsalz Hydrat, analytical standard
Phenylephrinhydrochlorid für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(R)-(−)-Phenylephrin -hydrochlorid, analytical standard
Sigma-Aldrich
Anti-NFκB p65 Antibody, CT, Upstate®, from rabbit