Merck
  • Recognition of riboflavin and the capsular polysaccharide of Haemophilus influenzae type b by antibodies generated to the haptenic epitope D-ribitol.
Recognition of riboflavin and the capsular polysaccharide of Haemophilus influenzae type b by antibodies generated to the haptenic epitope D-ribitol.
Glycoconjugate journal (2014-03-20)
G Ravi, Yeldur P Venkatesh
ZUSAMMENFASSUNG
D-Ribitol, a five-carbon sugar alcohol, is an important metabolite in the pentose phosphate pathway; it is an integral part of riboflavin (vitamin B2) and cell wall polysaccharides in most Gram-positive and a few Gram-negative bacteria. Antibodies specific to D-ribitol were generated in New Zealand white rabbits by using reductively aminated D-ribose-BSA conjugate as the immunogen. MALDI-TOF and amino group analyses of ribitol-BSA conjugate following 120 h reaction showed ~27-30 mol of ribitol conjugated per mole BSA. The presence of sugar alcohol in the conjugates was also confirmed by an increase in molecular mass and a positive periodic acid-Schiff staining in SDS-PAGE. Caprylic acid precipitation of rabbit serum followed by hapten affinity chromatography on ribitol-KLH-Sepharose CL-6B resulted in pure ribitol-specific antibodies (~45-50 μg/mL). The affinity constant of ribitol antibodies was found to be 2.9 × 10(7) M(-1) by non-competitive ELISA. Ribitol antibodies showed 100% specificity towards ribitol, ~800% cross-reactivity towards riboflavin, 10-15% cross-reactivity with sorbitol, xylitol and mannitol, and 5-7% cross-reactivity with L-arabinitol and meso-erythritol. The specificity of antibody to ribitol was further confirmed by its low cross-reactivity (0.4%) with lumichrome. Antibodies to D-ribitol recognized the purified capsular polysaccharide of Haemophilus influenzae type b, which could be specifically inhibited by ribitol. In conclusion, antibodies specific to D-ribitol have been generated and characterized, which have potential applications in the detection of free riboflavin and ribitol in biological samples, as well as identification of cell-surface macromolecules containing ribitol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumcyanoborhydrid, reagent grade, 95%
Sigma-Aldrich
Adenin, ≥99%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Thionylchlorid, ReagentPlus®, ≥99%
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Supelco
(−)-Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenin, BioReagent, suitable for cell culture
Sigma-Aldrich
Thionylchlorid, reagent grade, 97%
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Riboflavin 5′-Monophosphat Natriumsalz Hydrat, 70-79% (fluorimetric)
Sigma-Aldrich
Riboflavin 5′-monphosphate Natriumsalz Hydrat, ≥95% (HPLC)
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
Riboflavin 5′-Monophosphat Natriumsalz Hydrat, synthetic, ≥70% (HPLC)
Sigma-Aldrich
Riboflavin 5′-phosphate sodium salt hydrate, meets USP testing specifications
Sigma-Aldrich
Natriumcyanoborhydrid -Lösung, 1.0 M in THF
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumcyanoborhydrid -Lösung, 5.0 M in 1 M NaOH
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-(−)-Ribose, BioReagent, suitable for cell culture
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Riboflavin 5′-Monophosphat Natriumsalz Hydrat, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
Adenin, BioReagent, suitable for plant cell culture, ≥99%
Supelco
Adenin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lumichrom
Riboflavin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Adenin, European Pharmacopoeia (EP) Reference Standard