Merck
  • The effects and underlying mechanism of excessive iodide on excessive fluoride-induced thyroid cytotoxicity.
The effects and underlying mechanism of excessive iodide on excessive fluoride-induced thyroid cytotoxicity.
Environmental toxicology and pharmacology (2014-08-12)
Hongliang Liu, Qiang Zeng, Yushan Cui, Linyu Yu, Liang Zhao, Changchun Hou, Shun Zhang, Lei Zhang, Gang Fu, Yeming Liu, Chunyang Jiang, Xuemin Chen, Aiguo Wang
ZUSAMMENFASSUNG
In many regions, excessive fluoride and excessive iodide coexist in groundwater, which may lead to biphasic hazards to human thyroid. To explore fluoride-induced thyroid cytotoxicity and the mechanism underlying the effects of excessive iodide on fluoride-induced cytotoxicity, a thyroid cell line (Nthy-ori 3-1) was exposed to excessive fluoride and/or excessive iodide. Cell viability, lactate dehydrogenase (LDH) leakage, reactive oxygen species (ROS) formation, apoptosis, and the expression levels of inositol-requiring enzyme 1 (IRE1) pathway-related molecules were detected. Fluoride and/or iodide decreased cell viability and increased LDH leakage and apoptosis. ROS, the expression levels of glucose-regulated protein 78 (GRP78), IRE1, C/EBP homologous protein (CHOP), and spliced X-box-binding protein-1 (sXBP-1) were enhanced by fluoride or the combination of the two elements. Collectively, excessive fluoride and excessive iodide have detrimental influences on human thyroid cells. Furthermore, an antagonistic interaction between fluoride and excessive iodide exists, and cytotoxicity may be related to IRE1 pathway-induced apoptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumiodid, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumiodid, ReagentPlus®, 99%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Kaliumiodid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Kaliumiodid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Kaliumiodid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Kaliumiodid, ≥99.99% trace metals basis
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Kaliumiodid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumiodid, suitable for plant cell culture
Sigma-Aldrich
Kaliumiodid, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Kaliumiodid, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumiodid, AnhydroBeads, −10 mesh, 99.998% trace metals basis