Merck
  • Prevalence of groups A and C rotavirus antibodies in infants with biliary atresia and cholestatic controls.
Prevalence of groups A and C rotavirus antibodies in infants with biliary atresia and cholestatic controls.
The Journal of pediatrics (2014-12-03)
Maria Grazia Clemente, John T Patton, Robert Yolken, Peter F Whitington, Umesh D Parashar, Baoming Jiang, Trivellore Raghunathan, Kathleen B Schwarz
ZUSAMMENFASSUNG
To analyze the prevalence of acute asymptomatic group A and C rotavirus (RV-A and RV-C) infection in neonates with cholestasis. Participants were infants <180 days of age with cholestasis (serum direct or conjugated bilirubin >20% of total and ≥2 mg/dL) enrolled in the Childhood Liver Disease Research and Education Network during RV season (December-May). Forty infants with biliary atresia (BA), age 62 ± 29 days (range, 4.7-13 weeks) and 38 infants with cholestasis, age 67 ± 44 days (range, 3-15.8 weeks) were enrolled. At enrollment, RV-A IgM positivity rates did not differ between infants with BA (10%) vs those without (18%) (P = .349). RV-C IgM was positive in 0% of infants with BA vs 3% in those without BA (P = .49). RV-A IgG was lower in infants with BA: 51 ± 39 vs 56 ± 44 enzyme-linked immunoassay unit, P = .045 but this difference may lack biological relevance as maternal RV-A IgG titers were similar between groups. Infant RV-A IgM titers at 2-6 months follow-up increased markedly vs at presentation in both infants with BA (50 ± 30 vs 9 ± 9) and those without (43 ± 18 vs 16 ± 20 enzyme-linked immunoassay unit) (P < .0001), without differences between groups. RV-A infection in the first 6 months of life is common in infants with cholestasis of any cause. RV-A could have different pathogenetic effects by initiating different hepatic immune responses in infants with vs without BA or could lack pathogenetic significance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98.0% (NT)
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, tablet, 1 mg substrate per tablet
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Chlorwasserstoff–2-Propanol -Lösung, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
3,3′,5,5′-Tetramethylbenzidin, standard for GC