Merck
  • Thyroglobulin level in fine-needle aspirates for preoperative diagnosis of cervical lymph node metastasis in patients with papillary thyroid carcinoma: two different cutoff values according to serum thyroglobulin level.
Thyroglobulin level in fine-needle aspirates for preoperative diagnosis of cervical lymph node metastasis in patients with papillary thyroid carcinoma: two different cutoff values according to serum thyroglobulin level.
Thyroid : official journal of the American Thyroid Association (2015-01-22)
Min Ji Jeon, Won Gu Kim, Eun Kyung Jang, Yun Mi Choi, Yu-Mi Lee, Tae-Yon Sung, Jong Ho Yoon, Ki-Wook Chung, Suck Joon Hong, Jung Hwan Baek, Jeong Hyun Lee, Tae Yong Kim, Young Kee Shong, Won Bae Kim
ZUSAMMENFASSUNG
Measurement of thyroglobulin (Tg) in the washout fluid of fine-needle aspirates (FNA-Tg) is useful for diagnosis of lymph node (LN) metastasis in papillary thyroid carcinoma (PTC). However, the cutoff value of FNA-Tg in the preoperative state is not defined clearly. This study aimed to evaluate the optimal cutoff value of preoperative FNA-Tg according to serum Tg level. FNA-Tg was measured in 135 PTC patients (160 LNs) for preoperative diagnosis of cervical LN metastasis. Of the 160 LNs, 119 (74%) were surgically removed and 110 (69%) were diagnosed as malignant. When we adopted a FNA-Tg of 1.0 μg/L as the cutoff value, the sensitivity and specificity were 99% and 76%, respectively. FNA-Tg levels were correlated with serum Tg levels (Pearson's coefficient 0.42, p=0.002) and the FNA-Tg levels of 12 of the 50 benign LNs were above 1.0 μg/L. We classified the LNs into two groups according to serum Tg level regardless of anti-Tg antibody status: a low Tg group (≤1.0 μg/L, n=22, 14%) and a high Tg group (>1.0 μg/L, n=138, 86%). In the low Tg group, the sensitivity and specificity of the FNA-Tg cutoff value of 1.0 μg/L were 93% and 100%, respectively. In the high Tg group, the sensitivity and specificity of the FNA-Tg cutoff value of 19.0 μg/L were 93% and 100%, respectively. A Tg ratio (FNA-Tg level divided by serum Tg level) of 0.5 gave an improved diagnostic performance (sensitivity, 98%; specificity, 98%) in the high Tg group. FNA-Tg levels in the preoperative state are affected by serum Tg levels when they exceeded 1.0 μg/L. For the preoperative diagnosis of metastatic cervical LNs, it seems reasonable to employ different cutoff values of FNA-Tg depending on serum Tg levels. We propose the use of an optimal cutoff value of FNA-Tg of 1.0 μg/L in patients with low serum Tg levels and a Tg ratio of 0.5 in those with high serum Tg levels irrespective of thyroglobulin antibody status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-lactosamin, ≥98% (TLC)