Merck
  • The development of self-emulsifying oil-in-water emulsion adjuvant and an evaluation of the impact of droplet size on performance.
The development of self-emulsifying oil-in-water emulsion adjuvant and an evaluation of the impact of droplet size on performance.
Journal of pharmaceutical sciences (2015-01-21)
Ruchi R Shah, Stephanie Dodd, Mary Schaefer, Mildred Ugozzoli, Manmohan Singh, Gillis R Otten, Mansoor M Amiji, Derek T O'Hagan, Luis A Brito
ZUSAMMENFASSUNG
Microfluidization is an established technique for preparing emulsion adjuvant formulations for use in vaccines. Although this technique reproducibly yields high-quality stable emulsions, it is complex, expensive, and requires proprietary equipment. For this study, we developed a novel and simple low shear process to prepare stable reproducible emulsions without the use of any proprietary equipment. We found this process can produce a wide range of differently sized emulsions based on the modification of ratios of oil and surfactants. Using this process, we prepared a novel 20-nm-sized emulsion that was stable, reproducible, and showed adjuvant effects. During evaluation of this emulsion, we studied a range of emulsions with the same composition all sized below 200; 20, 90, and 160 nm in vivo and established a correlation between adjuvant size and immune responses. Our studies indicate that 160-nm-sized emulsions generate the strongest immune responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Squalen, ≥98%, liquid
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
SAFC
BIS-TRIS
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Supelco
Squalen, analytical standard
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri