Merck
A therapeutic approach to treat prostate cancer by targeting Nm23-H1/h-Prune interaction.
Naunyn-Schmiedeberg's archives of pharmacology (2014-08-21)
Marianeve Carotenuto, Pasqualino de Antonellis, Cristina Maria Chiarolla, Carmela Attanasio, Valentina Damiani, Iolanda Boffa, Nadia Aiese, Emilia Pedone, Benedetta Accordi, Giuseppe Basso, Luigi Navas, Ciro Imbimbo, Massimo Zollo
ZUSAMMENFASSUNG
Nm23-H1 is a metastasis suppressor gene whose overexpression is associated with both reduced cell motility in various cancers and increased metastatic potential in neuroblastomas, osteosarcomas, and hematological malignances. We previously reported that Nm23-H1 exerts tumor suppressor action in prostate cancer cells and that h-Prune, which is overexpressed in various tumor types, binds Nm23-H1. Moreover, blockage of the Nm23-H1/h-Prune interaction with a competitive permeable peptide (CPP) attenuates migration of breast and neuroblastoma cells. This series of events suggests that the Nm23-H1/h-Prune protein complex regulates cancer progression and that its specific impairment could be a new therapeutic strategy in oncology. We found that CPP leads to inhibition of the AKT/mTORv and NF-kBv signaling pathways and also activates apoptosis. To obtain a proof-of-concept of our hypothesis, we used a xenograft model of prostate cancer to evaluate whether impairment of this complex using CPP results in an anti-tumoral effect. Using a mouse orthotopic model with bioluminescent imaging, we show evidences that CPP reduces prostate cancer metastases formation. In conclusion, CPP being able to impair formation of the h-Prune/Nm23-H1 complex holds promise for the treatment of prostate cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamin
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Supelco
Glycerin, analytical standard
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland