- Nano-hydroxyapatite/polyacrylamide composite hydrogels with high mechanical strengths and cell adhesion properties.
Nano-hydroxyapatite/polyacrylamide composite hydrogels with high mechanical strengths and cell adhesion properties.
Colloids and surfaces. B, Biointerfaces (2014-12-04)
Zhiyong Li, Wenying Mi, Huiliang Wang, Yunlan Su, Changcheng He
PMID25465752
ZUSAMMENFASSUNG
Nano-hydroxyapatite/polyacrylamide composite hydrogels were successfully fabricated by physically mixing nano-hydroxyapatite (nHAp) particles into a peroxidized micelles initiated and cross-linked (pMIC) polyacrylamide (PAAm) hydrogel. The nanocomposite hydrogels exhibited excellent mechanical properties. The fracture tensile stresses of the gels were in the range of 0.21-0.86 MPa and the fracture tensile strains were up to 30 mm/mm, and the compressive strengths were up to 35.8 MPa. Meanwhile the introduction of nHAp endowed the composite hydrogels with good cell adhesion properties. This nHAp/PAAm nanocomposite hydrogel is expected to find potential applications in tissue engineering.