Merck
Gene pyramiding enhances durable blast disease resistance in rice.
Scientific reports (2015-01-15)
Shuichi Fukuoka, Norikuni Saka, Yuko Mizukami, Hironori Koga, Utako Yamanouchi, Yosuke Yoshioka, Nagao Hayashi, Kaworu Ebana, Ritsuko Mizobuchi, Masahiro Yano
ZUSAMMENFASSUNG
Effective control of blast, a devastating fungal disease of rice, would increase and stabilize worldwide food production. Resistance mediated by quantitative trait loci (QTLs), which usually have smaller individual effects than R-genes but confer broad-spectrum or non-race-specific resistance, is a promising alternative to less durable race-specific resistance for crop improvement, yet evidence that validates the impact of QTL combinations (pyramids) on the durability of plant disease resistance has been lacking. Here, we developed near-isogenic experimental lines representing all possible combinations of four QTL alleles from a durably resistant cultivar. These lines enabled us to evaluate the QTLs singly and in combination in a homogeneous genetic background. We present evidence that pyramiding QTL alleles, each controlling a different response to M. oryzae, confers strong, non-race-specific, environmentally stable resistance to blast disease. Our results suggest that this robust defence system provides durable resistance, thus avoiding an evolutionary "arms race" between a crop and its pathogen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salicylsäure, ACS reagent, ≥99.0%
Supelco
Salicylsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Salicylsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Salicylsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylen, ≥99.5%
Sigma-Aldrich
Salicylsäure, ≥99%, FG
Sigma-Aldrich
Salicylsäure, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylsäure, suitable for plant cell culture
Sigma-Aldrich
Salicylsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylsäure, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Salicylsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Salicylsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylsäure, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®