Merck
  • Multiwalled carbon nanotube-modified poly(D,L-lactide-co-glycolide) scaffolds for dendritic cell load.
Multiwalled carbon nanotube-modified poly(D,L-lactide-co-glycolide) scaffolds for dendritic cell load.
Journal of biomedical materials research. Part A (2014-06-10)
Yanzhu Yang, Sanyuan Shi, Qian Ding, Jian Chen, Jinliang Peng, Yuhong Xu
ZUSAMMENFASSUNG
Poly(D,L-lactide-co-glycolide) (PLGA) is widely used in a variety of tissue engineering and drug delivery applications due to its biodegradability and biocompatibility. But PLGA surfaces are usually hydrophobic which limited the loading and seeding capacities for cells, especially semiadherent immune cells. In this paper we described an attempt to improve the hydrophilicity and surface architecture for accommodating dendritic cells (DCs) that are widely used as professional antigen presenting cells in immune therapy of cancer and other diseases. The 3D porous PLGA scaffold was made by solvent casting/salt leaching of PLGA blended with surface functionalized multiwalled carbon nanotubes (F-MWCNTs). The incorporation and dispersion of F-MWCNT in the scaffold structures resulted in not only improved surface hydrophilicity but also nanoscale surface structure that would provide a preferable microenvironment for DCs attachment. We think such a scaffold material may be more desirable for immune cell delivery for immunotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
Thionylchlorid, ReagentPlus®, ≥99%
Supelco
Schwefelsäurekonzentrat, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Supelco
Salpetersäure-Konzentrat, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Thionylchlorid, reagent grade, 97%
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Supelco
Schwefelsäure, for the determination of nitrogen, ≥97.0%
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Schwefelsäure, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N