Merck
  • Critical contribution of MCL-1 in EMT-associated chemo-resistance in A549 non-small cell lung cancer.
Critical contribution of MCL-1 in EMT-associated chemo-resistance in A549 non-small cell lung cancer.
International journal of oncology (2015-02-04)
Masayoshi Toge, Satoru Yokoyama, Shinichiro Kato, Hiroaki Sakurai, Kazutaka Senda, Yoshinori Doki, Yoshihiro Hayakawa, Naoki Yoshimura, Ikuo Saiki
ZUSAMMENFASSUNG
Non-small cell lung cancer (NSCLC) is one of the leading causes of death in all lung cancer patients due to its metastatic spread. Even though cisplatin treatment after surgical resection of the primary tumor has been established as a standard chemotherapy for residual disease including metastatic spread, NSCLC often acquires a resistance against chemotherapy, and metastatic disease is often observed. Amongst many potential mechanisms, epithelial-to-mesenchymal transition (EMT) has been considered as an important process in acquiring both metastatic spread and chemo-resistance of NSCLC. In this study, we identified MCL-1 as a critical molecule for chemo-resistance in A549 cells associated with TGF-β-induced EMT. Importantly, downregulation of MCL-1 by siRNA or inhibition of MCL-1 with pan-BCL2 inhibitor to inhibit MCL-1 was able to overcome the EMT-associated chemo-resistance in A549 cells. Collectively, MCL-1 can be a new therapeutic target for overcoming EMT-associated chemo-resistance in NSCLC patients in the context of post-operative chemotherapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Sigma-Aldrich
trans-Platin(II)diammindichlorid
Cisplatin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard