Merck
  • Flow injection combined with ICP-MS for accurate high throughput analysis of elemental impurities in pharmaceutical products according to USP <232>/<233>.
Flow injection combined with ICP-MS for accurate high throughput analysis of elemental impurities in pharmaceutical products according to USP <232>/<233>.
Journal of pharmaceutical and biomedical analysis (2014-03-29)
Lisa Fischer, Barbara Zipfel, Gunda Koellensperger, Jessica Kovac, Susanne Bilz, Andrea Kunkel, Cornel Venzago, Stephan Hann
ZUSAMMENFASSUNG
New guidelines of the United States Pharmacopeia (USP), European Pharmacopeia (EP) and international organization (ICH, International Conference on Harmonization) regulating elemental impurity limits in pharmaceuticals seal the end of unspecific analysis of metal(oid)s as outlined in USP <231> and EP 2.4.8. Chapter USP <232> and EP 5.20 as well as drafts from ICH Q3D specify both daily doses and concentration limits of metallic impurities in pharmaceutical final products and in active pharmaceutical ingredients (API) and excipients. In chapters USP <233> and EP 2.4.20 method implementation, validation and quality control during the analytical process are described. By contrast with the--by now--applied methods, substance specific quantitative analysis features new basic requirements, further, significantly lower detection limits ask for the necessity of a general changeover of the methodology toward sensitive multi element analysis by ICP-AES and ICP-MS, respectively. A novel methodological approach based on flow injection analysis and ICP-SFMS/ICP-QMS for the quick and accurate analysis of Cd, Pb, As, Hg, Ir, Os, Pd, Pt, Rh, Ru, Cr, Mo, Ni, V, Cu, Mn, Fe and Zn in drug products by prior dilution, dissolution or microwave assisted closed vessel digestion according to the regulations is presented. In comparison to the acquisition of continuous signals, this method is advantageous with respect to the unprecedented high sample throughput due to a total analysis time of approximately 30s and the low sample consumption of below 50 μL, while meeting the strict USP demands on detection/quantification limits, precision and accuracy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Supelco
Salpetersäure-Konzentrat, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.