Merck
  • Enhancement of specificity of aldosterone measurement in human serum and plasma using 2D-LC-MS/MS and comparison with commercial immunoassays.
Enhancement of specificity of aldosterone measurement in human serum and plasma using 2D-LC-MS/MS and comparison with commercial immunoassays.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-24)
Julie A Ray, Mark M Kushnir, John Palmer, Seyed Sadjadi, Alan L Rockwood, A Wayne Meikle
ZUSAMMENFASSUNG
Accurate measurement of aldosterone is important for the standardized testing of primary hyperaldosteronism. Commercial immunoassays show substantial between-method variations resulting in significant clinical consequences. We developed a specific two dimensional (2D)-LC-MS/MS method for measuring aldosterone in human serum and plasma and compared it with three commercial immunoassays and an LC-MS/MS method. 250 μL samples, controls and calibrators spiked with d4-aldosterone were subjected to liquid-liquid extraction. The samples were analyzed using negative mode electrospray and 2D-LC followed by MS detection using an ABSciex 5500 mass spectrometer and compared with immunoassays of Siemens (Coat-A-Count), DiaSorin (CLIA-LIAISON), and IBL (ELISA). Data was acquired using multiple reaction-monitoring mode. LOQ and LOD of the method were 0.04 and 0.02 nmol/L respectively. The assay was linear up to 166 nmol/L. Inter and intra-assay imprecision at 0.13, 1.38 and 8.30 nmol/L were <10%. Interferences were absent and no differences were observed between serum and plasma matrices. Method recovery ranged from 95% to 113%. Ion suppression was not observed. Evaluated immunoassays showed positive biases ranging between 22% and 37% when compared with the developed method. We developed and validated an accurate method for measurement of aldosterone in human serum and plasma using 2D-LC-MS/MS which is suitable for clinical purposes. The method is faster than previously published LC-MS/MS methods, uses less sample, has adequate sensitivity while being able to preserve high specificity in a cost effective manner. Linearity of the assay makes it promising for urine and adrenal venous samples. Comparison with three commercial immunoassays demonstrates the advantages of the developed method.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
tert-Butylmethylether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
tert-Butylmethylether, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
tert-Butylmethylether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butylmethylether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aldosteron, ≥95% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Ammoniumacetat -Lösung, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammoniumacetat, BioUltra, ≥99.0%
Supelco
tert-Butylmethylether, analytical standard