Merck
  • Dried blood spot analysis of creatinine with LC-MS/MS in addition to immunosuppressants analysis.
Dried blood spot analysis of creatinine with LC-MS/MS in addition to immunosuppressants analysis.
Analytical and bioanalytical chemistry (2014-12-30)
Remco A Koster, Ben Greijdanus, Jan-Willem C Alffenaar, Daan J Touw
ZUSAMMENFASSUNG
In order to monitor creatinine levels or to adjust the dosage of renally excreted or nephrotoxic drugs, the analysis of creatinine in dried blood spots (DBS) could be a useful addition to DBS analysis. We developed a LC-MS/MS method for the analysis of creatinine in the same DBS extract that was used for the analysis of tacrolimus, sirolimus, everolimus, and cyclosporine A in transplant patients with the use of Whatman FTA DMPK-C cards. The method was validated using three different strategies: a seven-point calibration curve using the intercept of the calibration to correct for the natural presence of creatinine in reference samples, a one-point calibration curve at an extremely high concentration in order to diminish the contribution of the natural presence of creatinine, and the use of creatinine-[(2)H3] with an eight-point calibration curve. The validated range for creatinine was 120 to 480 μmol/L (seven-point calibration curve), 116 to 7000 μmol/L (1-point calibration curve), and 1.00 to 400.0 μmol/L for creatinine-[(2)H3] (eight-point calibration curve). The precision and accuracy results for all three validations showed a maximum CV of 14.0% and a maximum bias of -5.9%. Creatinine in DBS was found stable at ambient temperature and 32 °C for 1 week and at -20 °C for 29 weeks. Good correlations were observed between patient DBS samples and routine enzymatic plasma analysis and showed the capability of the DBS method to be used as an alternative for creatinine plasma measurement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar