Merck
  • Low in vitro permeability of the cyanotoxin microcystin-LR across a Caco-2 monolayer: with identification of the limiting factors using modelling.
Low in vitro permeability of the cyanotoxin microcystin-LR across a Caco-2 monolayer: with identification of the limiting factors using modelling.
Toxicon : official journal of the International Society on Toxinology (2014-08-12)
Jerome Henri, Antoine Huguet, Jean-Michel Delmas, Aurore Besson, Pascal Sanders, Valerie Fessard
ZUSAMMENFASSUNG
Microcystins (MCs) are toxins produced by several cyanobacteria species found worldwide. MC-LR is the most frequent. Here, we used the human Caco-2 cell line grown on semi-permeable filter supports as an in vitro model for determining MC-LR intestinal bidirectional transport. In this study, there was very low and time-dependent apparent permeability of MC-LR. To identify the limiting factors involved in the low permeability of MC-LR, a mathematical model was constructed to get physiologically relevant and informative parameters. The apical-to-basolateral transport was characterised by a rapid and substantial decrease in apical MC-LR concentrations (24-40% of the initial amount). In the basolateral compartment, the concentrations increased slowly after a lag time, but represented only a small fraction of the loaded concentrations (0.3-1.3%) after 24 h. This weak permeability was mainly due to a low clearance of efflux (from the cellular to the basolateral compartment) and effective secretion (from the cellular to the apical compartment). During the basolateral-to-apical transport, we observed a slow decrease in basolateral concentrations and a rapid increase in apical concentrations. In conclusion, modelling has the potential to highlight the key mechanisms involved in the complex kinetics of toxin transport.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard