Merck
  • Dynamic contrast-enhanced micro-computed tomography correlates with 3-dimensional fluorescence ultramicroscopy in antiangiogenic therapy of breast cancer xenografts.
Dynamic contrast-enhanced micro-computed tomography correlates with 3-dimensional fluorescence ultramicroscopy in antiangiogenic therapy of breast cancer xenografts.
Investigative radiology (2014-03-07)
Thomas Pöschinger, Anja Renner, Fabian Eisa, Michael Dobosz, Steffen Strobel, Thomas G Weber, Robert Brauweiler, Willi A Kalender, Werner Scheuer
ZUSAMMENFASSUNG
Dynamic contrast-enhanced (DCE) micro-computed tomography (micro-CT) has emerged as a valuable imaging tool to noninvasively obtain quantitative physiological biomarkers of drug effect in preclinical studies of antiangiogenic compounds. In this study, we explored the ability of DCE micro-CT to assess the antiangiogenic treatment response in breast cancer xenografts and correlated the results to the structural vessel response obtained from 3-dimensional (3D) fluorescence ultramicroscopy (UM). Two groups of tumor-bearing mice (KPL-4) underwent DCE micro-CT imaging using a fast preclinical dual-source micro-CT system (TomoScope Synergy Twin, CT Imaging GmbH, Erlangen, Germany). Mice were treated with either a monoclonal antibody against the vascular endothelial growth factor or an unspecific control antibody. Changes in vascular physiology were assessed measuring the mean value of the relative blood volume (rBV) and the permeability-surface area product (PS) in different tumor regions of interest (tumor center, tumor periphery, and total tumor tissue). Parametric maps of rBV were calculated of the tumor volume to assess the intratumoral vascular heterogeneity. Isotropic 3D UM vessel scans were performed from excised tumor tissue, and automated 3D segmentation algorithms were used to determine the microvessel density (MVD), relative vessel volume, and vessel diameters. In addition, the accumulation of coinjected fluorescence-labeled trastuzumab was quantified in the UM tissue scans to obtain an indirect measure of vessel permeability. Results of the DCE micro-CT were compared with corresponding results obtained by ex vivo UM. For validation, DCE micro-CT and UM parameters were compared with conventional histology and tumor volume. Examination of the parametric rBV maps revealed significantly different patterns of intratumoral blood supply between treated and control tumors. Whereas control tumors showed a characteristic vascular rim pattern with considerably elevated rBV values in the tumor periphery, treated tumors showed a widely homogeneous blood supply. Compared with UM, the physiological rBV maps showed excellent agreement with the spatial morphology of the intratumoral vascular architecture. Regional assessment of mean physiological values exhibited a significant decrease in rBV (P < 0.01) and PS (P < 0.05) in the tumor periphery after anti-vascular endothelial growth factor treatment. Structural validation with UM showed a significant reduction in reduction of relative vessel volume (rVV) (P < 0.01) and MVD (P < 0.01) in the corresponding tumor region. The reduction in rBV correlated well with the rVV (R = 0.73 for single values and R = 0.95 for mean values). Spatial maps of antibody penetration showed a significantly reduced antibody accumulation (P < 0.01) in the tumor tissue after treatment and agreed well with the physiological change of PS. Examination of vessel diameters revealed a size-dependent antiangiogenic treatment effect, which showed a significant reduction in MVD (P < 0.001) for vessels with diameters smaller than 25 μm. No treatment effect was observed by tumor volume. Noninvasive DCE micro-CT provides valuable physiological information of antiangiogenic drug effect in the intact animal and correlates with ex vivo structural analysis of 3D UM. The combined use of DCE micro-CT with UM constitutes a complementary imaging toolset that can help to enhance our understanding of antiangiogenic drug mechanisms of action in preclinical drug research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Benzylalkohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Benzylalkohol, ACS reagent, ≥99.0%
Supelco
Benzylalkohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Benzylalkohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Benzylalkohol, anhydrous, 99.8%
Sigma-Aldrich
Benzylalkohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzylbenzoat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Benzylalkohol, ≥99%, FCC, FG
Supelco
Benzylalkohol, analytical standard
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Benzylbenzoat, ≥99%, FCC, FG
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Benzylbenzoat, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzylbenzoat, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzylalkohol, natural, ≥98%, FG
Sigma-Aldrich
Benzylbenzoat, natural, ≥99%, FG
Benzylalkohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzylbenzoat, analytical standard