Merck
  • Determination of tacrine-6-ferulic acid in rat plasma by LC-MS/MS and its application to pharmacokinetics study.
Determination of tacrine-6-ferulic acid in rat plasma by LC-MS/MS and its application to pharmacokinetics study.
Biomedical chromatography : BMC (2014-04-08)
Xiuman Sun, Peiting Zhang, Rongbiao Pi, Yuting Zhou, Xuejiao Deng, Zhiyong Xie, Qiongfeng Liao
ZUSAMMENFASSUNG
Tacrine, as a drug for treating Alzheimer's disease (AD), has low efficacy owing to its single function and serious side effects. However, tacrine-6-ferulic acid (T6FA), the dimer which added ferulic acid to tacrine, has been found to be a promising multifunctional drug candidate for AD and much more potent and selective on acetylcholinesterase (AChE) than tacrine. The aim of the present work was to develop and validate an LC-MS/MS method with electrospray ionization for the quantification of T6FA in rat plasma using tacrine-3-ferulic acid (T3FA) as internal standard and to examine its application for pharmacokinetic study in rats. Following a single liquid-liquid extraction with ethyl acetate, chromatographic separation was achieved at 25 °C on a BDS Hypersil C18 column with a mobile phase composed of 1% formic acid and methonal (30:70, v/v) at a flow rate of 0.2 mL/min. Quantification was achieved by monitoring the selected ions at m/z 474.2 → 298.1 for T6FA and m/z 432.2 → 199.0 for T3FA. The method was validated to be rapid, specific, accurate and precise over the concentration range of 0.5-1000.0 ng/mL in rat samples. Furthermore, it was successfully applied for the pharmacokinetic measurement of T6FA with an oral administration at 40 mg/kg to rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Ethylacetat, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ethylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%