Merck
  • Protective immunity against Naegleria fowleri infection on mice immunized with the rNfa1 protein using mucosal adjuvants.
Protective immunity against Naegleria fowleri infection on mice immunized with the rNfa1 protein using mucosal adjuvants.
Parasitology research (2015-01-22)
Jinyoung Lee, Jong-Kyun Yoo, Hae-Jin Sohn, Hee-kyoung Kang, Daesik Kim, Ho-Joon Shin, Jong-Hyun Kim
ZUSAMMENFASSUNG
The free-living amoeba, Naegleria fowleri, causes a fatal disease called primary amoebic meningoencephalitis (PAM) in humans and experimental animals. Of the pathogenic mechanism of N. fowleri concerning host tissue invasion, the adherence of amoeba to hose cells is the most important. We previously cloned the nfa1 gene from N. fowleri. The protein displayed immunolocalization in the pseudopodia, especially the food-cups structure, and was related to the contact-dependent mechanism of the amoebic pathogenicity in N. fowleri infection. The cholera toxin B subunit (CTB) and Escherichia coli heat-labile enterotoxin B subunit (LTB) have been used as potent mucosal adjuvants via the parenteral route of immunization in most cases. In this study, to examine the effect of protective immunity of the Nfa1 protein for N. fowleri infection with enhancement by CTB or LTB adjuvants, intranasally immunized BALB/c mice were infected with N. fowleri trophozoites for the development of PAM. The mean time to death of mice immunized with the Nfa1 protein using LTB or CTB adjuvant was prolonged by 5 or 8 days in comparison with that of the control mice. In particular, the survival rate of mice immunized with Nfa1 plus CTB was 100% during the experimental period. The serum IgG levels were significantly increased in mice immunized with Nfa1 protein plus CTB or LTB adjuvants. These results suggest that the Nfa1 protein, with CTB or LTB adjuvants, induces strong protective immunity in mice with PAM due to N. fowleri infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid