Merck
Targetable micelleplex hydrogel for long-term, effective, and systemic siRNA delivery.
Biomaterials (2014-06-22)
Young-Min Kim, Soo-Chang Song
ZUSAMMENFASSUNG
We developed a targetable micelleplex hydrogel as a new efficient systemic siRNA delivery material that functions as a targetable gene carrier, and a hydrogel capable of controlled release to overcome drawbacks of multiple administrations of systemic siRNA carriers due to decreased fluctuation of them in the serum. The micelleplexes, complexes between polymeric micelles and siRNAs could turn into gel after subcutaneous injection and be slowly released from the gel. The released micelleplexes selectively accumulated in the tumor and showed anti-tumor effect due to gene silencing for an extended period of time with only one injection in anywhere in vivo model. Moreover, the duration of therapy can be controlled by adjusting the amount and properties of the hydrogel. Therefore, this micelleplex hydrogel is expected to be a new effective siRNA delivery material for systemic long-term gene silencing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
Ethanolamin, ≥99%
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Ethanolamin, ≥98%
Supelco
Folsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanolamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethanolamin, ACS reagent, ≥99.0%
USP
Folsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Folsäure, meets USP testing specifications
Sigma-Aldrich
Ethanolamin, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamin, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Ethanolamin, analytical standard
Trolamin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
Supelco
Aucubin, analytical standard
Folsäure, European Pharmacopoeia (EP) Reference Standard
Aucubin, primary reference standard
Sigma-Aldrich
MISSION® esiRNA, targeting human VEGFA
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vegfa