Merck
  • Oral toxicity evaluation of kefir-isolated Lactobacillus kefiranofaciens M1 in Sprague-Dawley rats.
Oral toxicity evaluation of kefir-isolated Lactobacillus kefiranofaciens M1 in Sprague-Dawley rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-05-21)
E E Owaga, M J Chen, W Y Chen, C W Chen, R H Hsieh
ZUSAMMENFASSUNG
Lactobacilli kefiranofaciens M1 has shown novel immunomodulation and anti-allergy probiotic attributes in cell and animal models. An acute oral toxicity assessment of L. kefiranofaciens M1 was evaluated in Sprague-Dawley rats. The rats were randomly assigned to four groups (12 rats/sex/group): the low dose group was orally gavaged with L. kefiranofaciens M1 at 3.0×10(8)cfu/kg bw while the medium dose and high dose groups received 9.0×10(9)cfu/kg bw and 1.8×10(10)cfu/kg bw, respectively, for 28days. The control group received phosphate buffer saline. The body weights were measured weekly while blood samples were collected for haematology and serum biochemistry tests. Histopathology of the organs (heart, liver, kidney, adrenal glands, spleen, ovary, testis), and urinalysis were conducted on study termination. The body weight gain of the L. kefiranofaciens M1 and control groups were comparable during the administration period. Overall, L. kefiranofaciens M1 did not induce adverse effects on haematology, serum biochemistry, and urinalysis parameters. Gross and microscopic histopathology of the organs revealed no toxicity effect of L. kefiranofaciens M1. In conclusion, 1.8×10(10)cfu/kg bw of L. kefiranofaciens M1 was considered as the no-observed-adverse-effect-level (NOAEL), which was the highest dose tested in the present study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Supelco
Diethylether, analytical standard
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C